2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%
2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%
2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%
2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%
Thermo Scientific Chemicals
2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%
CAS: 21679-14-1 | C10H12FN5O4 | 285.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ65562.031 g
Catalog number ALFJ65562.03
Price (MYR)
582.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
582.00
EA
Add to cart
Specifications
Chemical Name or Material2-Fluoro-9-beta-D-arabinofuranosyladenine
CAS21679-14-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
ColorWhite to Yellow