5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, 98%
Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, also known as bronidox, shows antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi. | CAS: 30007-47-7 | C4H6BrNO4 | 211.999 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ65443.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Bromo-5-nitro-1,3-dioxane
CAS30007-47-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth
Application
• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative
RUO – Research Use Only
General References:
  1. Ya. Legin, G. 2-Bromo-2-nitro-1,3-propanediol(Bronopol) and its derivatives: Synthesis, properties, and application (a review). Pharmaceutical Chemistry Journal. 1996, 30, (4), 273-284.
  2. Ghannoum, M.; Thomson, M.; Bowman, W.; Al-Khalil, S. Mode of action of the antimicrobial compound 5-Bromo-5-nitro-1,3-dioxane (Bronidox). Folia Microbiologica. 1986, 31, (1), 19-31.