Protease Inhibitor Cocktail I, Animal Free
Protease Inhibitor Cocktail I, Animal Free
Thermo Scientific Chemicals
Protease Inhibitor Cocktail I, Animal Free
Have Questions?
Catalog NumberQuantity
ALFJ65358.LQ1 Unit
Catalog number ALFJ65358.LQ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Unit
Request bulk or custom format
Specifications
ColorWhite
Product TypeProtease Inhibitor Cocktail
Recommended StorageStore at -20°C
SensitivityAir Sensitive
FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C