8-Aminonaphthalene-2-sulfonic acid, 95%
8-Aminonaphthalene-2-sulfonic acid, 95%
8-Aminonaphthalene-2-sulfonic acid, 95%
8-Aminonaphthalene-2-sulfonic acid, 95%
Thermo Scientific Chemicals
8-Aminonaphthalene-2-sulfonic acid, 95%
CAS: 119-28-8 | C10H9NO3S | 223.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ65304.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material8-Aminonaphthalene-2-sulfonic acid
CAS119-28-8
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c