5-Methyl-2'-deoxycytidine, 99%
5-Methyl-2'-deoxycytidine, 99%
5-Methyl-2'-deoxycytidine, 99%
5-Methyl-2'-deoxycytidine, 99%
Thermo Scientific Chemicals
5-Methyl-2'-deoxycytidine, 99%
CAS: 838-07-3 | C10H15N3O4 | 241.247 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ65209.031 g
Catalog number ALFJ65209.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Methyl-2'-deoxycytidine
CAS838-07-3
ColorWhite
Recommended StorageKeep cold
RTECS NumberHA3860000
View more
5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methyl-2′-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditio
RUO – Research Use Only
General References:
  1. Brian C. Froehler.; Daniel J. Ricca. Triple-helix formation by oligodeoxynucleotides containing the carbocyclic analogs of thymidine and 5-methyl-2'-deoxycytidine. J. Am. Chem. Soc. 1992, 114, (21), 8320-8322.
  2. Kuei-Ying Lin.; Robert J. Jones.; Mark Matteucci. Tricyclic 2'-Deoxycytidine Analogs: Syntheses and Incorporation into Oligodeoxynucleotides Which Have Enhanced Binding to Complementary RNA. J. Am. Chem. Soc. 1995, 117, (13), 3873-3874.