Cefoperazone sodium salt
Cefoperazone sodium salt
Cefoperazone sodium salt
Cefoperazone sodium salt
Thermo Scientific Chemicals
Cefoperazone sodium salt
Cefoperazone sodium salt, CAS # 62893-20-3, is the sodium salt form of cefoperazone. It is a semi-synthetic, broad-spectrum, and third-generation cephalosporin compound with antibiotic and bactericidal activities. | CAS: 62893-20-3 | C25H29N9NaO8S2 | 670.672 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ65185.031 g
Catalog number ALFJ65185.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCefoperazone sodium salt
TypeCefoperazone Sodium Salt
Physical FormPowder
CAS62893-20-3
Health Hazard 1H317-H334
View more
For studying the expression, binding, and inhibition of penicillin-binding proteins.Cefoperazone sodium salt is used in the study of drug-protein binding, expression and inhibition of penicillin-binding proteins during cell wall synthesis. It is as an antibacterial and antimicrobial agent useful for prevention of postoperative infection. It is used as an inhibitor of inactivation of alfa-antitrypsin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Cefoperazone sodium salt is a semi-synthetic, broad-spectrum, third-generation cephalosporin compound with antibiotic and bactericidal activities
• This compound can inhibit bacterial cell wall synthesis by inactivating penicillin-binding proteins and consequently interfering with the final transpeptidation step required for cross-linking peptidoglycan units, an important component of bacterial cell walls. This leads to a reduction in cell wall stability, causing cell lysis
Application
• Cefoperazone sodium salt exhibits activity against β-lactamase resistant microorganisms
• This compound is also used for studying the expression, binding, and inhibition of penicillin-binding proteins
• Cefoperazone is active in vitro against most gram-positive and gram-negative bacteria, including β-lactamase strains
• This compound has the ability to inhibit ∝-antitrypsin
RUO – Research Use Only
General References:
  1. Neu, H. The in vitro activity, human pharmacology, and clinical effectiveness of new β-lactam antibiotics. Annual Reviews of Pharmacology and Toxicology. 1982, 22, 599-642.
  2. Shahzad, M.; Abbas, A.; Rehman, R.; Mahmud, T.; Shamim, S. Comparative Analysis of Cefotaxime Sodium and Cefoperazone Sodium Drugs Activity and their Phytochemical Combinations Against Bacteria. Asian J. Chem. 2015, 27 (5), 1704-1706.
  3. Hajime, T.; Iwasaki, M.; Ogihara, H. Development of a Novel Chromogenic Medium for Improved Campylobacter Detection from Poultry Samples. J. Food Prot. 2015, 78 (9), 1750-1755.