N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
Thermo Scientific Chemicals
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
CAS: 21653-40-7 | C19H23ClN6O4 | 434.881 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Catalog number ALFJ64915.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialN-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
Melting Point223°C
CAS21653-40-7
ColorWhite to Yellow
Recommended StorageStore at -20°C
View more
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
Solubility
Soluble in DMSO at 50mg/ml
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Kayoko Yamaji.; Naotoshi Tsuji.; Takeharu Miyoshi.; M. Khyrul Islam.; Takeshi Hatta.; M. Abdul Alim.; M. Anisuzzaman.; Shiro Kushibiki.; Kozo Fujisaki. A salivary cystatin, HlSC-1, from the ixodid tick Haemaphysalis longicornis play roles in the blood-feeding processes. Parasitology Research. 2009, 106 (1), 61-68.
  2. Yoshiko Uesugi.; Jiro Arima.; Hirokazu Usuki.; Masaki Iwabuchi.; Tadashi Hatanaka. Two bacterial collagenolytic serine proteases have different topological specificities. Biochimica et Biophysica Acta (BBA) - Proteins and Proteomics. 2008, 1784(4), 716-726.