2'-Amino-3'-methoxyflavone, 99%
2'-Amino-3'-methoxyflavone, 99%
2'-Amino-3'-methoxyflavone, 99%
2'-Amino-3'-methoxyflavone, 99%
Thermo Scientific Chemicals
2'-Amino-3'-methoxyflavone, 99%
A MAP kinase inhibitor | CAS: 167869-21-8 | C16H13NO3 | 267.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ64485.MA10 mg
Catalog number ALFJ64485.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Chemical Name or Material2'-Amino-3'-methoxyflavone
CAS167869-21-8
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
A MAP kinase inhibitor