Penicillin G potassium salt
Penicillin G potassium salt
Penicillin G potassium salt
Penicillin G potassium salt
Thermo Scientific Chemicals
Penicillin G potassium salt
Inhibits bacterial cell wall synthesis | CAS: 113-98-4 | C33H36ClKN4O4S2 | 691.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ63901.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPenicillin G potassium salt
TypePenicillin G Potassium Salt
Physical FormPowder
CAS113-98-4
Health Hazard 1H317-H334
View more
Penicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Penicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.
Solubility
Soluble in water (100 mg/ml), methanol, ethanol (sparingly), and alcohol. Insoluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Li-Li Du, Qi Wu,; Chun-Xiu Chen,; Bo-Kai Liu,; Xian-Fu Lin. A two-step, one-pot enzymatic synthesis of ampicillin from penicillin G potassium salt . Journal of Molecular Catalysis B: Enzymatic. 2009, 58 (1-4) , 208-211.
  2. Junhong Qian,; Rong Guo, Xia Guo. The hydrolysis of penicillin-G potassium salt in the O/W microemulsions with different charges. Colloids and Surfaces A: Physicochemical and Engineering Aspects. 2003, 215(1-3), 253-262.