Brilliant Blue G soln., Ready-to-Use
Brilliant Blue G soln., Ready-to-Use
Brilliant Blue G soln., Ready-to-Use
Brilliant Blue G soln., Ready-to-Use
Thermo Scientific Chemicals
Brilliant Blue G soln., Ready-to-Use
CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALFJ63797.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBrilliant Blue G soln.
Name NoteReady-to-Use
CAS6104-58-1
Health Hazard 1H500
Health Hazard 2GHS H Statement
H311-H331-H370-H226
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Flammable liquid and vapor.