Diacerein
Diacerein
Diacerein
Thermo Scientific Chemicals
Diacerein
Interleukin-1 inhibitor | CAS: 13739-02-1 | C19H12O8 | 368.30 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63775.031 g
Catalog number ALFJ63775.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiacerein
CAS13739-02-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Interleukin-1 inhibitor