L-Cysteine, Cell Culture Reagent
L-Cysteine, Cell Culture Reagent
L-Cysteine, Cell Culture Reagent
L-Cysteine, Cell Culture Reagent
Thermo Scientific Chemicals
L-Cysteine, Cell Culture Reagent
CAS: 52-90-4 | C3H7NO2S | 121.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ63745.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Cysteine
Melting Point∽220°C (decomposition)
CAS52-90-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.
Solubility
Soluble in water (10 mg/ml), 1N HCl (50 mg/ml), and ethanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from air.
RUO – Research Use Only
General References:
  1. Yukio Kurita.; Walter Gordy. Electron Spin Resonance in a Gamma-Irradiated Single Crystal of L-Cystine Dihydrochloride.J. Chem. Phys. 1961, 34 (1),282 .
  2. Limin Zheng,; Robert H. White,; Valerie L. Cash,; Dennis R. Dean. Mechanism for the Desulfurization of L-Cysteine Catalyzed by the nifS Gene Product. Journal name. 1994, 33 (15), 4714-4720.