Proteinase K, Ready-to-Use Soln.
Proteinase K, Ready-to-Use Soln.
Actual product may vary
Proteinase K, Ready-to-Use Soln.
Proteinase K, Ready-to-Use Soln.
Proteinase K, Ready-to-Use Soln.
A stable and highly reactive serine protease; used for protein and nucleic acid isolation
Have Questions?
Catalog NumberQuantity
ALFJ63710.AA5 mL
Catalog number ALFJ63710.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS39450-01-6
ColorClear, Colorless
Formula Weight28.9 kd
For Use With (Application)A stable and highly reactive serine protease; used for protein and nucleic acid isolation
Product TypeDNA Extraction Reagent
Quantity5 mL
FormLiquid,28.9 kDa
Unit SizeEA
Contents & Storage
-30°C to -10°C