Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Isosorbide mononitrate, 98+%
Thermo Scientific Chemicals
Isosorbide mononitrate, 98+%
A coronary vasodilator that does not require hepatic biotransformation | CAS: 16051-77-7 | C6H9NO6 | 191.139 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63674.065 g
Catalog number ALFJ63674.06
Price (MYR)
1,370.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,370.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsosorbide mononitrate
CAS16051-77-7
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
A coronary vasodilator that does not require hepatic biotransformation