Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Thermo Scientific Chemicals
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63669.AK250 mL
Catalog number ALFJ63669.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium acetate
Name NoteRNAse free
CAS127-09-3
pH4.5
Recommended StorageAmbient temperatures
View more
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only