Sucrose, 1.2M aq. soln., Thermo Scientific™
Sucrose, 1.2M aq. soln., Thermo Scientific™
Sucrose, 1.2M aq. soln., Thermo Scientific™
Sucrose, 1.2M aq. soln., Thermo Scientific™
Sucrose, 1.2M aq. soln., Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFJ63662.AP500 mL
Catalog number ALFJ63662.AP
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSucrose
CAS57-50-1
Recommended StorageKeep cold
TSCAYes
Molecular FormulaC12H22O11
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.