Chlorpromazine hydrochloride, 98+%
Chlorpromazine hydrochloride, 98+%
Chlorpromazine hydrochloride, 98+%
Chlorpromazine hydrochloride, 98+%
Thermo Scientific Chemicals
Chlorpromazine hydrochloride, 98+%
CAS: 69-09-0 | C17H20Cl2N2S | 355.321 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Catalog number ALFJ63659.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChlorpromazine hydrochloride
CAS69-09-0
Health Hazard 1H301-H330
Health Hazard 2GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.
Health Hazard 3P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
View more
Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorpromazine hydrochloride is used as a substitute for benzidine, o-dianisidine and o-tolidine in the determination of microquantities of hemoglobin and peroxidase. Chlorpromazine, Hydrochloride is used in cell signaling and neuroscience research.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Yongnian Ni.; Li Wang.; Serge Kokot. Voltammetric determination of chlorpromazine hydrochloride and promethazine hydrochloride with the use of multivariate calibration.Analytica Chimica Acta.2001, (1), 159-168.
  2. Wenbing Shi.; Jidong Yang.; Yuming Huang. Ion-pair complex-based solvent extraction combined with chemiluminescence determination of chlorpromazine hydrochloride with luminol in reverse micelles.Journal of Pharmaceutical and Biomedical Analysis.2001, (1), 197-203.