Finasteride
Finasteride
Finasteride
Thermo Scientific Chemicals
Finasteride
Inhibitor of steroid 5-a-reductase | CAS: 98319-26-7 | C23H36N2O2 | 372.553 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
1 g
5 g
Catalog number ALFJ63454.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialFinasteride
CAS98319-26-7
Health Hazard 1H302-H360D
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Peter Boyle.; Lawrence Gould, A.; Claus G. Roehrborn. Prostate volume predicts outcome of treatment of benign prostatic hyperplasia with finasteride: Meta-analysis of randomized clinical trials *.Urology.1996, (3), 398-405.
  2. Matuszewski, B.K.; Constanzer, M.L.; Chavez-Eng, C.M. Matrix Effect in Quantitative LC/MS/MS Analyses of Biological Fluids: A Method for Determination of Finasteride in Human Plasma at Picogram Per Milliliter Concentrations.Anal. Chem.1998, (5), 882-889.