Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Thermo Scientific Chemicals
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ63394.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium n-dodecyl sulfate (SDS)
ColorColorless
CAS151-21-3
Health Hazard 1H315-H318
Physical FormLiquid
View more
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.
Solubility
Miscible with ethanol.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhao, M.; Yuan, J.; Zheng, L. Spontaneous formation of vesicles by N-dodecyl-N-methylpyrrolidinium bromide (C 12 MPB) ionic liquid and sodium dodecyl sulfate (SDS) in aqueous solution. Colloids Surf., A 2012, 407, 116-120.
  2. Chen, S.; Duhamel, J.; Peng, B.; Zaman, M.; Tam, K. C. Interactions between a Series of Pyrene End-Labeled Poly (ethylene oxide) s and Sodium Dodecyl Sulfate in Aqueous Solution Probed by Fluorescence. Langmuir 2014, 30, (44), 13164-13175.