Tween 20 washing buffer, 1% in PBS (10X)
Tween 20 washing buffer, 1% in PBS (10X)
Thermo Scientific Chemicals
Tween 20 washing buffer, 1% in PBS (10X)
Have Questions?
Catalog NumberQuantity
ALFJ63314.K21 L
Catalog number ALFJ63314.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Product TypeBuffer
Quantity1 L
FormViscous Liquid
Concentration1% in PBS (10X)
Unit SizeEA
Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Keep cold