Fexofenadine hydrochloride
Fexofenadine hydrochloride
Fexofenadine hydrochloride
Fexofenadine hydrochloride
Thermo Scientific Chemicals
Fexofenadine hydrochloride
H-1 receptor antagonist | CAS: 153439-40-8 | C32H40ClNO4 | 538.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Catalog number ALFJ63262.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialFexofenadine hydrochloride
CAS153439-40-8
Melting Point148°C to 150°C
ColorWhite
Recommended StorageStore at -20°C
View more
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.
Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Harold S. Nelson.; Robert Reynolds.; Jolene Mason. Fexofenadine HCl is safe and effective for treatment of chronic idiopathic urticaria.Annals of Allergy, Asthma & Immunology.2000, 84, (5), 517-522.
  2. Fran Ridout.; Ziba Shamsi.; Robert Meadows.; Sigurd Johnson.; Ian Hindmarch. A single-center, randomized, double-blind, placebo-controlled, crossover investigation of the effects of fexofenadine hydrochloride 180 mg alone and with alcohol, with hydroxyzine hydrochloride 50 mg as a positive internal control, on aspects of cognitive and psychomotor function related to driving a car.Clinical Therapeutics.2003, 25, (5), 1518-1538.
  3. Fran Ridout.; Ziba Shamsi.; Robert Meadows.; Sigurd Johnson.; Ian Hindmarch. A single-center, randomized, double-blind, placebo-controlled, crossover investigation of the effects of fexofenadine hydrochloride 180 mg alone and with alcohol, with hydroxyzine hydrochloride 50 mg as a positive internal control, on aspects of cognitive and psychomotor function related to driving a car.Clinical Therapeutics.2003, 25, (5), 1518-1538.