Dulcitol, 99%
Dulcitol, 99%
Dulcitol, 99%
Thermo Scientific Chemicals
Dulcitol, 99%
Useful reagent in bacteriology | CAS: 608-66-2 | C6H14O6 | 182.172 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63206.30250 g
Catalog number ALFJ63206.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDulcitol
CAS608-66-2
Melting Point187°C to 191°C
ColorWhite
Recommended StorageAmbient temperatures
View more
Useful reagent in bacteriology