Sodium citrate, 0.5M buffer soln., pH 5.5
Sodium citrate, 0.5M buffer soln., pH 5.5
Thermo Scientific Chemicals
Sodium citrate, 0.5M buffer soln., pH 5.5
Have Questions?
Catalog NumberQuantity
ALFJ63199.AK250 mL
Catalog number ALFJ63199.AK
Price (MYR)
634.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
634.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium citrate buffer soln.
Health Hazard 1Warning
ColorColorless
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
View more
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 
Solubility
Soluble in water.
Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.
RUO – Research Use Only
General References:
  1. CJ Quaife.; SD Findley.; JC Erickson.; GJ Froelick. Induction of a new metallothionein isoform (MT-IV) occurs during differentiation of stratified squamous epithelia. Biochemistry. 1994, 33,(23), 7250-7259.
  2. M Cappello.; S Keshav.; C Prince.; DP Jewell.; S Gordon. Detection of mRNAs for macrophage products in inflammatory bowel disease by in situ hybridization. Gut . 1992, 33,(9), 1214-1219.