Famotidine, 98+%
Famotidine, 98+%
Famotidine, 98+%
Thermo Scientific Chemicals
Famotidine, 98+%
A competitive histamine H2 receptor antagonist | CAS: 76824-35-6 | C8H15N7O2S3 | 337.435 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFJ63165.06
Price (MYR)
1,545.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,545.00
EA
Add to cart
Specifications
Chemical Name or MaterialFamotidine
CAS76824-35-6
Melting Point166°C to 168°C
ColorWhite
Recommended StorageAmbient temperatures
View more
Famotidine, is commonly used in the treatment of peptic ulcer disease and gastroesophageal reflux disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. RP Walt.; MJS Langman.; J Cottrell.; SG Mann. Continuous intravenous famotidine for haemorrhage from peptic ulcer. The Lancet. 1992340 (8827), 1058-1062.
  2. M Jaimini.; AC Rana.; YS Tanwar. Formulation and evaluation of famotidine floating tablets. Current drug delivery. 20074 (1), 51-55.