Demeclocycline hydrochloride
Demeclocycline hydrochloride
Demeclocycline hydrochloride
Demeclocycline hydrochloride
Thermo Scientific Chemicals
Demeclocycline hydrochloride
CAS: 64-73-3 | C21H22Cl2N2O8 | 501.313 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63102.031 g
Catalog number ALFJ63102.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDemeclocycline hydrochloride
CAS64-73-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Mahrous, M.S.; Abdel-Khalek, M.M. Spectrophotometric determination of phenothiazines, tetracyclines and chloramphenicol with sodium cobaltinitrite. Talanta. 1984, 31, (4), 289-291.
  2. Santos, M.D.F.; Vermeersch, H.; Remon, J.P.; Schelkens, M.; De Backer, P.; Ducatelle, R.; Haesebrouck, F. Validation of a high-performance liquid chromatographic method for the determination of doxycycline in turkey plasma. Journal of Chromatography B: Biomedical Sciences and Applications. 1996, 682, (2), 301-308.