Vitamin A palmitate, 1.7 M.I.U./g
Vitamin A palmitate, 1.7 M.I.U./g
Vitamin A palmitate, 1.7 M.I.U./g
Vitamin A palmitate, 1.7 M.I.U./g
Thermo Scientific Chemicals
Vitamin A palmitate, 1.7 M.I.U./g
Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. | CAS: 79-81-2 | C36H60O2 | 524.874 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ63022.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVitamin A palmitate
CAS79-81-2
Health Hazard 1H360
Health Hazard 3P201-P202-P281-P308+P313-P501c
Melting Point28°C to 29°C
View more
Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. Other applications include as pharmaceutical product which is used for manufacturing aqueous solutions using solubilizing agents such as cremophor. It is also used in the fortification of fatty foods such as margarine, spreads, oils, milk and dairy products. It is used in cosmetics for the preparation of creams and lotions.