Chelerythrine chloride, 99+%
Chelerythrine chloride, 99+%
Chelerythrine chloride, 99+%
Chelerythrine chloride, 99+%
Thermo Scientific Chemicals
Chelerythrine chloride, 99+%
CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62906.MA10 mg
Catalog number ALFJ62906.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialChelerythrine chloride
CAS3895-92-9
Health Hazard 1H302+H312+H332-H315-H319
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells
RUO – Research Use Only