L-Asparagine monohydrate, Cell Culture Reagent
L-Asparagine monohydrate, Cell Culture Reagent
L-Asparagine monohydrate, Cell Culture Reagent
L-Asparagine monohydrate, Cell Culture Reagent
Thermo Scientific Chemicals
L-Asparagine monohydrate, Cell Culture Reagent
CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62869.22100 g
Catalog number ALFJ62869.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Asparagine monohydrate
Melting Point215°C to 217°C
CAS5794-13-8
ColorWhite
Recommended StorageAmbient temperatures
View more
It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. EE Haley.; GA Fischer.; AD Welc. The requirement for L-asparagine of mouse leukemia cells L5178Y in culture. Cancer research. 1961, 21 (1), 532-34.
  2. HJ Son.; HG Kim.; KK Kim.; HS Kim.; YG Kim. Increased production of bacterial cellulose by Acetobacter sp. V6 in synthetic media under shaking culture conditions. Bioresource Technology. 1961, 86 (3), 215-219.