Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free
Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free
Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free
Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62848.AK250 mL
Catalog number ALFJ62848.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTris(hydroxymethyl)aminomethane hydrochloride
ColorColorless
Name NoteRNAse free
CAS1185-53-1
Recommended StorageAmbient temperatures
View more
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.