Doxepin hydrochloride
Doxepin hydrochloride
Doxepin hydrochloride
Doxepin hydrochloride
Thermo Scientific Chemicals
Doxepin hydrochloride
Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist | CAS: 1229-29-4 | C19H22ClNO | 315.841 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62579.031 g
Catalog number ALFJ62579.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDoxepin hydrochloride
CAS1229-29-4
Health Hazard 1H301-H361
Health Hazard 2GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Health Hazard 3P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
View more
It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Tran VT, et al. [3H]doxepin interactions with histamine H1-receptors and other sites in guinea pig and rat brain homogenates.Eur J Pharmacol.,1981,70(4), 501-9.
  2. Luchins DJ. Review of clinical and animal studies comparing the cardiovascular effects of doxepin and other tricyclic antidepressants.Am J Psychiatry.,1983,140(8), 1006-9.