Flumazenil, 98%
Flumazenil, 98%
Flumazenil, 98%
Thermo Scientific Chemicals
Flumazenil, 98%
Benzodiazepine antagonist | CAS: 78755-81-4 | C15H14FN3O3 | 303.293 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62537.MC100 mg
Catalog number ALFJ62537.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialFlumazenil
CAS78755-81-4
ColorWhite
Recommended StorageKeep cold
RTECS NumberNI2922170
View more
Benzodiazepine antagonist