Tetraethylammonium chloride
Tetraethylammonium chloride
Tetraethylammonium chloride
Tetraethylammonium chloride
Thermo Scientific Chemicals
Tetraethylammonium chloride
Phase transfer catalyst | CAS: 56-34-8 | C8H20ClN | 165.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ62507.22
Price (MYR)
789.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
789.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetraethylammonium chloride
CAS56-34-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.
Solubility
Soluble in water, ethanol, chloroform, or acetone
Notes
Hygroscopic, Incompatible with strong oxidizing agents. Protect from moisture.  Keep container tightly closed in a dry and well-ventilated place. Handle under argon. Store in cool place.
RUO – Research Use Only
General References:
  1. Wei AD et al. Nomenclature and molecular relationships of calcium-activated potassium channels. Pharmacol Rev. 2005, 57, 463-72.
  2. Niu et al. Vasorelaxant effect of taurine is diminished by tetraethylammonium in rat isolated arteries. Eur.J.Pharmacol. 2008, 580, (1-2),169-74.