Penicillin V potassium salt
Penicillin V potassium salt
Penicillin V potassium salt
Penicillin V potassium salt
Thermo Scientific Chemicals
Penicillin V potassium salt
Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62442.0910 g
Catalog number ALFJ62442.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPenicillin V potassium salt
TypePenicillin V Potassium Salt
Physical FormPowder
CAS132-98-9
Health Hazard 1H302-H317-H334
View more
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.
Solubility
Soluble in water, dimethyl sulfoxide, and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
RUO – Research Use Only
General References:
  1. Li-Li Du, Qi Wu.; Chun-Xiu Chen.; Bo-Kai Liu.; Xian-Fu Lin. A two-step, one-pot enzymatic synthesis of ampicillin from penicillin G potassium salt.J. Mol. Catal. B: Enzym. 2009, 58 (1-4),208-211 .
  2. John C. Sheehan.; Kenneth R. Henery-Logan. The Total Synthesis of Penicillin V.J. Am. Chem. Soc. 1959, 81 (12),3089-3094 .