Triolein, tech.
Triolein, tech.
Triolein, tech.
Thermo Scientific Chemicals
Triolein, tech.
CAS: 122-32-7 | C57H104O6 | 885.453 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ62419.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTriolein
CAS122-32-7
ColorYellow
Recommended StorageStore at -20°C
Density0.91
View more
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.
Solubility
Soluble in chlorofom, ether, and carbon tetrachloride
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C
RUO – Research Use Only
General References:
  1. A Salis.; M Pinna.; M Monduzzi.; V Solinas. Biodiesel production from triolein and short chain alcohols through biocatalysis. Journal of Biotechnology. 2005, 119, (3), 291-299.
  2. F Mori.; JC Lim.; OG Raney.; CM Elsik.; CA Miller. Phase behavior, dynamic contacting and detergency in systems containing triolein and nonionic surfactants. Colloids and surfaces. 1989, 40, (1), 323-345.