Glycerol, Cell Culture Grade
Glycerol, Cell Culture Grade
Glycerol, Cell Culture Grade
Glycerol, Cell Culture Grade
Glycerol, Cell Culture Grade
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62399.AP500 mL
Catalog number ALFJ62399.AP
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
Melting Point18°C
Recommended StorageAmbient temperatures
Density1.26
View more
Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel's sample well.
Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component. Glycerol acts as a cryoprotectant and prevents formation of ice crystals and prevents cell damage. Glycerol is useful in a variety of applications, glycerol is used as a solvent, plasticizer and humectant.
In polyacrylamide gel electrophoresis, glycerol is used in sample preparation and gel formation. At a concentration of 5-10%, glycerol is used to increase the density of a sample so that it will layer at the bottom of a sample well. Additional uses include as an aid in casting gradient gels, a protein stabilizer and storage buffer component.