Triton∣r X-100 lysis buffer, pH 7.4
Triton∣r X-100 lysis buffer, pH 7.4
Thermo Scientific Chemicals
Triton∣r X-100 lysis buffer, pH 7.4
Have Questions?
Catalog NumberQuantity
ALFJ62289.AK250mL
Catalog number ALFJ62289.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250mL
Request bulk or custom format
Specifications
Quantity250mL
FormatLiquid
Recommended StorageKeep cold
Product TypeCell Lysis Reagent
pH7.4
View more
It is widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions. Triton X-100 is a commonly used detergent in laboratories. Triton X-100 is widely used to lyse cells to extract protein or organelles, or to permeabilize the membranes of living cells. Apart from laboratory use, Triton X-100 can be found in several types of cleaning compounds,[8] ranging from heavy-duty industrial products to gentle detergents. It is also a popular ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol. It is a good micellar catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2° to 8°C