Aloin
Aloin
Aloin
Thermo Scientific Chemicals
Aloin
Aloe exudant, used as a stimulant-laxative | CAS: 1415-73-2 | C21H22O9 | 418.40 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62153.MC100 mg
Catalog number ALFJ62153.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialAloin
CAS1415-73-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Aloe exudant, used as a stimulant-laxative