Sodium phosphate, 0.2M buffer soln., pH 7.4
Sodium phosphate, 0.2M buffer soln., pH 7.4
Thermo Scientific Chemicals
Sodium phosphate, 0.2M buffer soln., pH 7.4
Have Questions?
Catalog NumberQuantity
ALFJ62152.AP500 mL
Catalog number ALFJ62152.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate buffer soln.
Recommended StorageAmbient temperatures
Concentration0.2 M
TSCAYes
pH7.4
View more
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Incompatible with strong acids.
RUO – Research Use Only
General References:
  1. Tillmanns, E.; Baur, W. H. Stoichiometry of trisodium orthophosphate dodecahydrate. Inorg. Chem. 1970, 9 (8), 1957-1958.
  2. Tillmanns, E.; Baur, W. H. Influence of Coffee Genotype on Bioactive Compounds and the in Vitro Capacity To Scavenge Reactive Oxygen and Nitrogen Species. J. Agric. Food Chem. 2015, 63 (19), 4815-4826.