MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
Thermo Scientific Chemicals
MES-SDS running buffer (20X)
Have Questions?
Catalog NumberQuantity
ALFJ62138.K21 L
Catalog number ALFJ62138.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Recommended StorageAmbient temperatures
Concentration20X
Physical FormLiquid
Quantity1 L
Unit SizeEA
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.
Solubility
Miscible with water.
Notes
This buffer contains: 1M MES, 1M Tris base,2% SDS and 20mM EDTA at pH 7.3. Store at room temperature. Keep away from light. Incompatible with strong oxidizing agent and acids.
RUO – Research Use Only
General References:
  1. Costaa, J.; Ansaria, P.; Mafrab, I.; Oliveirab, M. B. P.P.; Baumgartner, S. Development of a sandwich ELISA-type system for the detection and quantification of hazelnut in model chocolates. Food Chem. 2015, 173, 257-265.
  2. Juvonen, K. R.; Macierzanka, A.; Lille, M. E.; Laaksonen, D. E.; Mykkänen, H. M.; Niskanen, L. K.; Pihlajamäki, J.; Mäkeläa, K. A.; Millsa, C. E. N.; Mackiea, A. R.; Malcolma, P.; Herziga, K. H.; Poutanena, K. S.; Karhunena, L. J. Cross-linking of sodium caseinate-structured emulsion with transglutaminase alters postprandial metabolic and appetite responses in healthy young individuals. Br. J. Nutr. 2015, 114 (03), 418-429.