Ofloxacin
Ofloxacin
Ofloxacin
Thermo Scientific Chemicals
Ofloxacin
Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity | CAS: 82419-36-1 | C18H20FN3O4 | 361.373 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62080.065 g
Catalog number ALFJ62080.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOfloxacin
TypeOfloxacin
Physical FormCrystalline Powder
CAS82419-36-1
Melting Point270°C to 275°C
View more
Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity