Lansoprazole, 98+%
Lansoprazole, 98+%
Lansoprazole, 98+%
Thermo Scientific Chemicals
Lansoprazole, 98+%
A proton pump inhibitor | CAS: 103577-45-3 | C16H14F3N3O2S | 369.362 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62008.031 g
Catalog number ALFJ62008.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLansoprazole
CAS103577-45-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
A proton pump inhibitor