Carbenicillin disodium salt
Carbenicillin disodium salt
Carbenicillin disodium salt
Carbenicillin disodium salt
Thermo Scientific Chemicals
Carbenicillin disodium salt
Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities. | CAS: 4800-94-6 | C17H16N2Na2O6S | 422.36 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61949.031 g
Catalog number ALFJ61949.03
Price (MYR)
716.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
716.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbenicillin disodium salt
TypeCarbenicillin Disodium Salt
Physical FormPowder
CAS4800-94-6
Health Hazard 1H317-H334-H335
View more
Carbenicillin disodium salt is a water-soluble antibiotic which is effective against gram-negative bacteria.Carbenicillin disodium salt is widely utilized as an antibiotic which is used to control bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It has been involved in the study of the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium
Application
  • Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
RUO – Research Use Only
General References:
  1. Cole, M. R.; Hobden, J. A.; Warner, I. M. Recycling Antibiotics into GUMBOS: A New Combination Strategy to Combat Multi-Drug-Resistant Bacteria. Molecules 2015, 20 (4), 6466-6487.
  2. Sonopo, M. S.; Venter, K.; Boyle, G.; Winks, S.; Painter, B. M.; Zeevaart, J. R. Carbon-14 radiolabeling and in vivo biodistribution of a potential anti-TB compound. J. Labelled Comp. Radiopharm. 2015, 58 (2), 23-29.