Paraformaldehyde, 4% in PBS
Paraformaldehyde, 4% in PBS
Paraformaldehyde, 4% in PBS
Paraformaldehyde, 4% in PBS
Paraformaldehyde, 4% in PBS
Thermo Scientific Chemicals
Paraformaldehyde, 4% in PBS
Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane, 500mL, CHEBI:16842, H317-H318-H351 | CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ61899.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde
Physical FormLiquid
CAS30525-89-4
Health Hazard 1Serious eye damage/eye irritation (category 1), Carcinogenicity (category 2), Skin sensitisation (category 1)
Health Hazard 2Causes serious eye damage, Suspected of causing cancer, May cause an allergic skin reaction
View more
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Rafi, M. A.; Rao, H. Z.; Luzi, P.; Luddi, A.; Curtis, M. T.; Wenger, D. A. Intravenous injection of AAVrh10-GALC after the neonatal period in twitcher mice results in significant expression in the central and peripheral nervous systems and improvement of clinical features. Mol. Genet. Metab. 2015, 114 (3), 459-466.
  2. de Couto, G.; Liu, W.; Tseliou, E.; Sun, B.; Makkar, N.; Kanazawa, H.; Arditi, M.; Marbán, E. Macrophages mediate cardioprotective cellular postconditioning in acute myocardial infarction. J. Clin. Invest. 2015, 125 (8), 3147-3162.