Lithium chloride, 1M aq. soln.
Lithium chloride, 1M aq. soln.
Lithium chloride, 1M aq. soln.
Lithium chloride, 1M aq. soln.
Thermo Scientific Chemicals
Lithium chloride, 1M aq. soln.
CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number ALFJ61895.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium chloride
CAS7447-41-8
ColorColorless
Recommended StorageAmbient temperatures
Physical FormLiquid