Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Thermo Scientific Chemicals
Ammonium peroxydisulfate, Electrophoresis Grade
CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ61856.22
Price (MYR)
521.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
521.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium peroxydisulfate
Melting Point120°C (decomposition)
CAS7727-54-0
ColorWhite
Health Hazard 1H272-H302-H315-H317-H319-H334-H335