Sodium pyruvate, Cell Culture Grade
Sodium pyruvate, Cell Culture Grade
Sodium pyruvate, Cell Culture Grade
Sodium pyruvate, Cell Culture Grade
Thermo Scientific Chemicals
Sodium pyruvate, Cell Culture Grade
CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
Catalog number ALFJ61840.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium pyruvate
CAS113-24-6
ColorWhite to Yellow
Recommended StorageAmbient temperatures
Physical FormPowder