Tolnaftate
Tolnaftate
Tolnaftate
Thermo Scientific Chemicals
Tolnaftate
Antifungal agent | CAS: 2398-96-1 | C19H17NOS | 307.411 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFJ61834.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTolnaftate
TypeTolnaftate
Physical FormPowder
CAS2398-96-1
Melting Point110°C to 113°C
View more
Tolnaftate is used to study the mechanism of ergosterol (sterol) biosynthesis at the rate-limiting step of squalene epoxidase inhibition. Tolnaftate is clinically used to treat cutaneous infections such as athlete?s foot, jock itch, and ringworm1. It also has been used as a comparator compound for antifungal efficiency studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only
General References:
  1. Craig Knox et. al. DrugBank 3.0: a comprehensive resource for omics research on drugs. Nucleic Acids Research. 2011, 39, D1035-D1041.
  2. J Pannu et. al. NB-002, a novel nanoemulsion with broad antifungal activity against dermatophytes, other filamentous fungi, and Candida albicans. Antimicrobial Agents and Chemotherapy. 2011, 53, (8), 3273-3279.