(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ61745.MF50 mg
Catalog number ALFJ61745.MF
Price (MYR)
804.00
EA
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (MYR)
804.00
EA
Add to cart
Specifications
Chemical Name or Material(-)-Epigallocatechin gallate
CAS989-51-5
Health Hazard 1H312
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P280-P302+P352-P312-P363-P501c
View more
Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).
Solubility
Soluble in ethanol, dimethyl formamide, water.