Prazosin hydrochloride
Prazosin hydrochloride
Prazosin hydrochloride
Prazosin hydrochloride
Thermo Scientific Chemicals
Prazosin hydrochloride
An alpha adrenoceptor antagonist | CAS: 19237-84-4 | C19H22ClN5O4 | 419.87 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61712.MD250 mg
Catalog number ALFJ61712.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialPrazosin hydrochloride
CAS19237-84-4
Health Hazard 1H302-H315-H319-H335-H361
Health Hazard 2GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin
Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Longxiao Liu.; Suyan Zhu. Preparation and characterization of inclusion complexes of prazosin hydrochloride with β-cyclodextrin and hydroxypropyl-β-cyclodextrin. Journal of Pharmaceutical and Biomedical Analysis. 2006, 40,(1), 122-127.
  2. Longxiao Liu.; Suyan Zhu. A study on the supramolecular structure of inclusion complex of β-cyclodextrin with prazosin hydrochloride. Carbohydrate Polymers. 2007, 68,(3), 472-476.