Luminol and oxidizing solutions (each bottle)
Luminol and oxidizing solutions (each bottle)
Thermo Scientific Chemicals
Luminol and oxidizing solutions (each bottle)
For chemiluminecent detection of HRP-based Western blotting systems
Have Questions?
Catalog NumberQuantity
ALFJ61462.AK250 mL
Catalog number ALFJ61462.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialLuminol and oxidizing solutions (each bottle)
Recommended StorageKeep cold
TSCAYes
Solubility InformationIt is soluble in most polar organic solvents but is insoluble in water.
Quantity250 mL
View more
Used by biologists in cellular assays for the detection of copper, iron, and cyanides, as well as of specific proteins by western blot. For chemiluminecent HRP-based detection of western blotting systems. Microestimation of glucose and glucose oxidase using enzyme-induced chemiluminescence6,7 - Inhibitor of poly ADP Ribose Synthase8 - Demonstrations of the phenomenon of chemiluminescence, using different sensitizers to produce different colors of light.